Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168335.1 | NP_001161807.1 | 5 | 688 | ||
NM_002396.4 | NP_002387.1 | 5 | 688 | |||
GenBank mRNA | AK313391.1 | - | 5 | 584 | ||
BC000147.1 | - | 5 | 630 | 2900677 | ||
BI559761.1 | - | 5 | 656 | |||
M55905.1 | - | 5 | 518 |