Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166434.2 | NP_001159906.1 | 7 | 813 | ||
NM_001166435.2 | NP_001159907.1 | 5 | 544 | |||
NM_001166436.2 | NP_001159908.1 | 5 | 430 | |||
NM_002215.3 | NP_002206.2 | 9 | 974 | |||
GenBank mRNA | AK292750.1 | - | 9 | 944 | ||
AK298398.1 | - | 9 | 973 | |||
AK298423.1 | - | 9 | 966 | |||
AK298455.1 | - | 7 | 799 | |||
AK303110.1 | - | 5 | 430 | |||
AK303156.1 | - | 5 | 544 | |||
AK314198.1 | - | 9 | 944 | |||
BC069464.1 | - | 9 | 968 | 7262464 | ||
X16260.1 | - | 7 | 723 | |||
X63652.1 | - | 9 | 968 |