Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304441.1 | NP_001291370.1 | 11 | 1846 | ||
NM_001304442.1 | NP_001291371.1 | 11 | 1843 | |||
NM_002424.2 | NP_002415.1 | 10 | 1755 | |||
XM_011542834.2 | XP_011541136.1 | 11 | 1835 | |||
XM_011542835.2 | XP_011541137.1 | 12 | 1922 | |||
XM_011542836.2 | XP_011541138.1 | 10 | 1681 | |||
XM_017017771.1 | XP_016873260.1 | 11 | 1712 | |||
GenBank mRNA | AK292659.1 | - | 10 | 1755 | ||
AK303388.1 | - | 11 | 1846 | |||
BC144094.1 | - | 11 | 1805 | |||
BG213446.1 | - | 2 | 469 | |||
BG218770.1 | - | 1 | 341 | |||
J05556.1 | - | 10 | 1727 |