Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002428.3 | NP_002419.1 | 3, 4 | 1219 | ||
GenBank mRNA | BC006412.1 | - | 3, 4 | 339 | ||
BC036495.1 | - | 3, 4 | 1225 | 5263145 | ||
BC055428.1 | - | 3, 4 | 1220 | 4811572 | ||
BT006715.1 | - | 1, 2 | 119 | |||
D86331.1 | - | 2, 3 | 157 | |||
Z48482.1 | - | 3, 4 | 482 |