Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002489.3 | NP_002480.1 | 4 | 432 | ||
GenBank mRNA | AF201077.1 | - | 1 | 356 | ||
AK024837.1 | - | 4 | 381 | |||
AK130950.1 | - | 4 | 372 | |||
BC101794.1 | - | 4 | 356 | 8069300 | ||
BC101796.1 | - | 4 | 356 | 8069302 | ||
BC105295.1 | - | 4 | 403 | 6739636 | ||
BG623521.1 | - | 433 | ||||
U94586.1 | - | 4 | 393 |