Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199987.1 | NP_001186916.1 | 3 | 457 | ||
NM_002493.4 | NP_002484.1 | 4 | 550 | |||
NM_182739.2 | NP_877416.1 | 3 | 505 | |||
GenBank mRNA | AF035840.1 | - | 4 | 494 | ||
AF067167.1 | - | 4 | 523 | |||
AK092649.1 | - | 1 | 1942 | |||
AK289702.1 | - | 4 | 504 | |||
AW160372.1 | - | 3 | 388 | |||
BC009801.1 | - | 4 | 529 | 4334414 | ||
BC016708.1 | - | 4 | 564 | |||
BC029247.1 | - | 4 | 495 | 5171627 | ||
BG220924.1 | - | 2 | 162 | |||
BI669356.1 | - | 3 | 484 | |||
BP208846.1 | - | 4 | 548 |