Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199987.1 | NP_001186916.1 | 3 | 553 | ||
NM_002493.4 | NP_002484.1 | 4 | 646 | |||
NM_182739.2 | NP_877416.1 | 3 | 601 | |||
GenBank mRNA | AK289702.1 | - | 4 | 600 | ||
AW160372.1 | - | 3 | 484 | |||
BG220924.1 | - | 2 | 258 | |||
BI669356.1 | - | 3 | 580 |