Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318051.1 | NP_001304980.1 | 2 | 173 | ||
NM_002495.3 | NP_002486.1 | 2 | 173 | |||
NR_134473.1 | 2 | 173 | ||||
NR_134474.1 | 2 | 173 | ||||
NR_134475.1 | 3 | 297 | ||||
XM_017009491.1 | XP_016864980.1 | 2 | 196 | |||
GenBank mRNA | AF020351.1 | - | 2 | 151 | ||
AK307973.1 | - | 2 | 151 | |||
BC005270.1 | - | 2 | 167 | 3681312 | ||
BU145342.1 | - | 2 | 147 | |||
BU954271.1 | - | 2 | 165 | |||
CD692133.1 | - | 3 | 313 | |||
DA609203.1 | - | 2 | 173 | |||
DB489661.1 | - | 2 | 172 | |||
KU178226.1 | - | 2 | 143 | |||
KU178227.1 | - | 2 | 143 |