Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002496.3 | NP_002487.1 | 5 | 367 | ||
XM_005274013.1 | XP_005274070.1 | 5 | 464 | |||
XM_005274014.2 | XP_005274071.1 | 5 | 491 | |||
XM_011545053.2 | XP_011543355.1 | 5 | 402 | |||
GenBank mRNA | AK002110.1 | - | 3 | 955 | ||
AK223114.1 | - | 5 | 357 | |||
AK302449.1 | - | 5 | 367 | |||
AK314546.1 | - | 5 | 292 | |||
BC119754.2 | - | 5 | 296 | 40119290 | ||
DB498707.1 | - | 5 | 353 | |||
U65579.1 | - | 5 | 290 |