Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242361.1 | NP_001229290.1 | 3 | 388 | ||
NM_002525.2 | NP_002516.2 | 3 | 993 | |||
XM_011541522.1 | XP_011539824.1 | 4 | 982 | |||
XM_011541525.2 | XP_011539827.1 | 3 | 420 | |||
GenBank mRNA | AY360265.1 | - | 3 | 364 | ||
DB084774.1 | - | 3 | 386 | |||
X93207.1 | - | 3 | 806 |