Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101662.1 | NP_001095132.1 | 21 | 2655 | ||
NM_001242361.1 | NP_001229290.1 | 23 | 2254 | |||
NM_002525.2 | NP_002516.2 | 23 | 2859 | |||
XM_005270903.1 | XP_005270960.1 | 21 | 2002 | |||
XM_011541522.1 | XP_011539824.1 | 24 | 2848 | |||
XM_011541523.1 | XP_011539825.1 | 21 | 2219 | |||
XM_011541525.2 | XP_011539827.1 | 23 | 2317 | |||
XM_017001375.1 | XP_016856864.1 | 21 | 2079 | |||
GenBank mRNA | AK055516.1 | - | 21 | 1999 | ||
AK299271.1 | - | 21 | 2102 | |||
AK302616.1 | - | 21 | 2079 | |||
AK312318.1 | - | 21 | 2467 | |||
AY049784.1 | - | 21 | 2370 | |||
AY360265.1 | - | 23 | 2230 | |||
BC008775.2 | - | 21 | 2429 | 3138712 | ||
U64898.1 | - | 21 | 2427 | |||
X93207.1 | - | 23 | 2672 | |||
X93209.1 | - | 21 | 2468 |