Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002581.4 | NP_002572.2 | 9 | 3272 | ||
XM_006717129.2 | XP_006717192.1 | 7 | 1346 | |||
XM_017014784.1 | XP_016870273.1 | 9 | 3312 | |||
GenBank mRNA | AK300128.1 | - | 7 | 1499 | ||
AK300129.1 | - | 3 | 203 | |||
BC078657.1 | - | 9 | 2902 | 30340336 | ||
BU158714.1 | - | 3 | 391 | |||
X68280.1 | - | 9 | 2663 |