Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270481.1 | NP_001257410.1 | 3 | 267 | ||
NM_002798.2 | NP_002789.1 | 3 | 267 | |||
GenBank mRNA | AK311768.1 | - | 3 | 230 | ||
BC000835.2 | - | 3 | 234 | 3456009 | ||
BF671774.1 | - | 3 | 268 | |||
BM553399.1 | - | 3 | 214 | |||
BQ691763.1 | - | 3 | 249 | |||
BT009809.1 | - | 3 | 216 | |||
BU848746.1 | - | 3 | 199 | |||
CR457065.1 | - | 3 | 216 | |||
D29012.1 | - | 3 | 225 | |||
X61971.1 | - | 1 | 39 |