Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199771.1 | NP_001186700.1 | 2 | 429 | ||
NM_002905.3 | NP_002896.2 | 2 | 424 | |||
GenBank mRNA | AK293313.1 | - | 2 | 380 | ||
AK307157.1 | - | 2 | 251 | |||
BC028298.1 | - | 2 | 383 | 4942311 | ||
BI765016.1 | - | 2 | 335 | |||
BP381700.1 | - | 2 | 424 | |||
U43559.1 | - | 2 | 317 | |||
U89717.1 | - | 2 | 354 |