Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNN1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130413.3 | NP_001123885.2 | 18 | 2952 | ||
NR_037668.2 | 17 | 2858 | ||||
XM_011541899.2 | XP_011540201.1 | 18 | 3622 | |||
XM_011541901.2 | XP_011540203.1 | 19 | 3144 | |||
XM_011541902.2 | XP_011540204.1 | 19 | 3120 | |||
XM_011541905.2 | XP_011540207.1 | 18 | 3048 | |||
XM_011541906.2 | XP_011540208.1 | 15 | 3382 | |||
XM_011541908.2 | XP_011540210.1 | 18 | 3319 | |||
XM_011541920.2 | XP_011540222.1 | 17 | 3142 | |||
XM_011541925.2 | XP_011540227.1 | 16 | 2701 | |||
XM_011541929.2 | XP_011540231.1 | 14 | 3510 | |||
XM_011541932.2 | XP_011540234.1 | 15 | 2607 | |||
XM_011541933.2 | XP_011540235.1 | 15 | 2573 | |||
XM_017002037.1 | XP_016857526.1 | 17 | 3619 | |||
XM_017002038.1 | XP_016857527.1 | 16 | 3472 | |||
XM_017002039.1 | XP_016857528.1 | 18 | 2997 | |||
XM_017002040.1 | XP_016857529.1 | 18 | 2859 | |||
XM_017002041.1 | XP_016857530.1 | 19 | 2847 | |||
XM_017002042.1 | XP_016857531.1 | 18 | 2838 | |||
XM_017002043.1 | XP_016857532.1 | 18 | 2814 | |||
XM_017002044.1 | XP_016857533.1 | 17 | 2763 | |||
GenBank mRNA | AK093239.1 | - | 15 | 2582 | ||
AK093372.1 | - | 12 | 1707 | |||
AK127357.1 | - | 15 | 3378 | |||
AK131558.1 | - | 19 | 3278 |