Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002259.4 | NP_002250.1 | 7 | 1063 | ||
NM_007328.3 | NP_015567.1 | 6 | 1009 | |||
NM_213657.2 | NP_998822.1 | 7 | 1183 | |||
NM_213658.2 | NP_998823.1 | 8 | 1237 | |||
GenBank mRNA | AF461812.1 | - | 7 | 1039 | ||
BC012550.1 | - | 7 | 1187 | 4042168 | ||
BC053840.1 | - | 8 | 1006 | 5484921 | ||
X54867.1 | - | 7 | 1013 | |||
X54868.1 | - | 6 | 959 |