Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003363.3 | NP_003354.2 | 9 | 1147 | ||
NM_199443.2 | NP_955475.1 | 8 | 1006 | |||
GenBank mRNA | AF017305.1 | - | 9 | 1071 | ||
AF017306.1 | - | 8 | 927 | |||
AK127055.1 | - | 3 | 1583 | |||
AK222725.1 | - | 8 | 936 | |||
AK223292.1 | - | 9 | 1077 | |||
AK291795.1 | - | 9 | 1092 | |||
BC125130.1 | - | 9 | 1118 | 40119090 | ||
BC125131.1 | - | 9 | 1115 | 40119091 | ||
U20657.1 | - | 9 | 1071 |