Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003725.3 | NP_003716.2 | 2 | 298 | ||
XM_005269207.1 | XP_005269264.1 | 3 | 447 | |||
XM_005269208.1 | XP_005269265.1 | 3 | 451 | |||
XM_005269209.1 | XP_005269266.1 | 3 | 438 | |||
XM_006719672.1 | XP_006719735.1 | 3 | 446 | |||
XM_011538925.1 | XP_011537227.1 | 3 | 441 | |||
XM_011538926.1 | XP_011537228.1 | 3 | 431 | |||
XM_011538927.1 | XP_011537229.1 | 3 | 422 | |||
GenBank mRNA | AF016509.1 | - | 1 | 82 | ||
AF223225.1 | - | 2 | 169 | |||
AK313423.1 | - | 2 | 208 | |||
BC020710.1 | - | 2 | 192 | 4700726 | ||
DA086202.1 | - | 3 | 431 | |||
U89281.1 | - | 2 | 187 |