Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161504.1 | NP_001154976.1 | 15 | 3228 | ||
NM_001319218.1 | NP_001306147.1 | 14 | 3215 | |||
NM_003748.3 | NP_003739.2 | 15 | 3368 | |||
NM_170726.2 | NP_733844.1 | 16 | 2355 | |||
GenBank mRNA | BC007581.1 | - | 15 | 3098 | 3139944 | |
BC023600.2 | - | 15 | 3109 | 4548787 | ||
BX649081.1 | - | 2 | 2471 | |||
FJ462711.1 | - | 14 | 2953 | |||
U24266.1 | - | 15 | 3121 | |||
U24267.1 | - | 16 | 2110 |