Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003793.3 | NP_003784.2 | 3 | 589 | ||
XM_011545328.2 | XP_011543630.1 | 3 | 339 | |||
GenBank mRNA | AF071748.1 | - | 1 | 153 | ||
AF071749.1 | - | 1 | 55 | |||
AF088886.2 | - | 3 | 589 | |||
AF136279.1 | - | 3 | 588 | |||
AJ007331.1 | - | 3 | 544 | |||
AK313657.1 | - | 3 | 557 | |||
AL137742.1 | - | 1 | 20 | |||
BC011682.2 | - | 3 | 583 | 3535532 | ||
BC036451.1 | - | 3 | 564 | 4830274 | ||
CR541680.1 | - | 1 | 61 |