Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003816.2 | NP_003807.1 | 22 | 2738 | ||
NR_027638.1 | 21 | 2596 | ||||
NR_027639.1 | 20 | 2551 | ||||
NR_027878.1 | 21 | 2632 | ||||
XM_011544682.2 | XP_011542984.1 | 22 | 2988 | |||
XM_017013942.1 | XP_016869431.1 | 22 | 2681 | |||
GenBank mRNA | BC126406.1 | - | 21 | 2556 | 8992122 | |
BC143923.1 | - | 21 | 2592 | 9052441 | ||
BC143924.1 | - | 20 | 2511 | |||
D14665.2 | - | 18 | 2313 | |||
U41766.1 | - | 22 | 2738 |