Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003940.2 | NP_003931.2 | 21 | 2711 | ||
XM_011513269.1 | XP_011511571.1 | 20 | 2631 | |||
XM_017007425.1 | XP_016862914.1 | 21 | 2555 | |||
XM_017007426.1 | XP_016862915.1 | 20 | 2522 | |||
XM_017007427.1 | XP_016862916.1 | 18 | 2242 | |||
GenBank mRNA | AK290338.1 | - | 21 | 2658 | ||
AK302404.1 | - | 21 | 2520 | |||
BC016146.1 | - | 21 | 2664 | 3910925 | ||
U75362.1 | - | 21 | 2724 |