Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004104.4 | NP_004095.4 | 8451 | |||
GenBank mRNA | BC007267.1 | - | 3593 | |||
BC007909.1 | - | 2866 | ||||
BC014634.2 | - | 1862 | ||||
BC063242.1 | - | 8451 | 6172538 | |||
S80437.1 | - | 2207 |