Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP8B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004391.2 | NP_004382.2 | 1 | 884 | ||
GenBank mRNA | AF090318.1 | - | 1 | 443 | ||
AK310784.1 | - | 1 | 641 | |||
AK315330.1 | - | 1 | 623 | |||
BC067434.1 | - | 1 | 592 | 7002161 | ||
BC067441.1 | - | 1 | 592 | 7002145 | ||
BC067442.1 | - | 1 | 592 | 7002146 | ||
BC067444.1 | - | 1 | 592 | 7002148 |