Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004519.3 | NP_004510.1 | 1, 2 | 611 | ||
GenBank mRNA | AB208890.1 | - | 1, 2 | 164 | ||
AF033347.1 | - | 1, 2 | 245 | |||
BC128576.1 | - | 2, 3 | 354 |