Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204824.1 | NP_001191753.1 | 10 | 1070 | ||
NM_004519.3 | NP_004510.1 | 10 | 1619 | |||
XM_005250914.3 | XP_005250971.1 | 7 | 694 | |||
XM_006716555.3 | XP_006716618.1 | 8 | 880 | |||
XM_011517026.2 | XP_011515328.1 | 10 | 4153 | |||
XM_017013400.1 | XP_016868889.1 | 10 | 11125 | |||
GenBank mRNA | AB208890.1 | - | 10 | 1172 | ||
AF033347.1 | - | 10 | 1253 | |||
AK296293.1 | - | 10 | 1070 | |||
BC128576.1 | - | 11 | 1362 |