Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143970.1 | NP_001137442.1 | 3 | 929 | ||
NM_001143971.1 | NP_001137443.1 | 3 | 853 | |||
NM_004824.3 | NP_004815.3 | 3 | 1080 | |||
NR_026590.1 | 5 | 1035 | ||||
GenBank mRNA | AF081258.1 | - | 5 | 1035 | ||
AF081259.1 | - | 5 | 1035 | |||
AK291601.1 | - | 4 | 843 | |||
AK296985.1 | - | 3 | 929 | |||
BC061516.1 | - | 2 | 150 | |||
BC108725.1 | - | 3 | 1091 | 6140263 | ||
BC119682.2 | - | 2 | 76 | 40116323 | ||
BG766689.1 | - | 2 | 305 |