Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206802.2 | NP_001193731.1 | 3 | 572 | ||
NM_004881.4 | NP_004872.2 | 3 | 1456 | |||
NM_147184.3 | NP_671713.1 | 4 | 1057 | |||
XM_005264650.2 | XP_005264707.1 | 4 | 929 | |||
XM_006712150.2 | XP_006712213.1 | 4 | 1057 | |||
GenBank mRNA | AF010309.1 | - | 3 | 1094 | ||
AK223382.1 | - | 3 | 1293 | |||
AK297579.1 | - | 3 | 897 | |||
AY371700.1 | - | 3 | 567 | |||
BC000474.2 | - | 4 | 627 | 2961571 | ||
BC018819.2 | - | 4 | 627 | |||
BT007149.1 | - | 3 | 567 |