Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289023.2 | NP_001275952.2 | 8 | 933 | ||
NM_001289025.2 | NP_001275954.2 | 9 | 984 | |||
NM_001289026.2 | NP_001275955.2 | 9 | 935 | |||
NM_001289027.2 | NP_001275956.2 | 8 | 902 | |||
NM_002287.5 | NP_002278.2 | 9 | 987 | |||
NM_021706.4 | NP_068352.2 | 8 | 936 | |||
NR_110279.2 | 9 | 893 | ||||
NR_110280.2 | 9 | 1228 | ||||
GenBank mRNA | AF013249.1 | - | 9 | 745 | ||
AF109683.1 | - | 8 | 710 | |||
AF251509.2 | - | 8 | 626 | |||
AF251510.2 | - | 8 | 768 | |||
AK093559.1 | - | 4 | 952 | |||
AK097869.1 | - | 8 | 868 | |||
AK290415.1 | - | 9 | 801 | |||
AK303261.1 | - | 9 | 831 | |||
AK309895.1 | - | 6 | 886 | |||
AK315419.1 | - | 9 | 833 | |||
BC027899.1 | - | 9 | 737 | 5228732 | ||
BI756388.1 | - | 9 | 649 | |||
CR542051.1 | - | 9 | 677 |