Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004995.3 | NP_004986.1 | 9, 10 | 1702 | ||
GenBank mRNA | AK291325.1 | - | 9, 10 | 1644 | ||
AK312657.1 | - | 9, 10 | 1650 | |||
BC064803.1 | - | 9, 10 | 1636 | 6046773 | ||
D26512.1 | - | 9, 10 | 1528 | |||
HE978355.1 | - | 9, 10 | 1651 | |||
U41078.1 | - | 9, 10 | 1417 | |||
X83535.1 | - | 9, 10 | 1536 | |||
X90925.1 | - | 9, 10 | 1529 | |||
Z48481.1 | - | 9, 10 | 1530 |