Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319214.1 | NP_001306143.1 | 7 | 991 | ||
NM_005040.3 | NP_005031.1 | 8 | 1353 | |||
NM_199418.3 | NP_955450.2 | 9 | 1416 | |||
XM_005274093.1 | XP_005274150.1 | 8 | 1083 | |||
GenBank mRNA | AB451270.1 | - | 8 | 1152 | ||
AB451397.1 | - | 8 | 1152 | |||
AK090847.1 | - | 2 | 1649 | |||
AK091786.1 | - | 9 | 1243 | |||
AK302386.1 | - | 7 | 942 | |||
AK312919.1 | - | 8 | 1174 | |||
BC001500.2 | - | 8 | 1171 | 3510081 | ||
L13977.1 | - | 8 | 1181 |