Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207053.1 | NP_001193982.1 | 5 | 1332 | ||
NM_001243126.1 | NP_001230055.1 | 5 | 1315 | |||
NM_005046.3 | NP_005037.1 | 6 | 1463 | |||
NM_139277.2 | NP_644806.1 | 6 | 1293 | |||
GenBank mRNA | AF411214.1 | - | 6 | 1437 | ||
AK295313.1 | - | 5 | 1311 |