Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128610.2 | NP_001122082.1 | 5 | 747 | ||
NM_001283049.1 | NP_001269978.1 | 3 | 376 | |||
NM_005154.4 | NP_005145.3 | 5 | 607 | |||
XM_006720761.3 | XP_006720824.1 | 6 | 927 | |||
XM_006720762.3 | XP_006720825.1 | 6 | 927 | |||
XM_011522193.2 | XP_011520495.1 | 6 | 764 | |||
XM_017022718.1 | XP_016878207.1 | 5 | 602 | |||
XM_017022719.1 | XP_016878208.1 | 5 | 719 | |||
XM_017022720.1 | XP_016878209.1 | 6 | 764 | |||
XM_017022721.1 | XP_016878210.1 | 6 | 664 | |||
XM_017022722.1 | XP_016878211.1 | 4 | 587 | |||
GenBank mRNA | AK292400.1 | - | 5 | 602 | ||
AK296480.1 | - | 3 | 366 | |||
BC015545.1 | - | 6 | 881 | |||
BC026954.1 | - | 5 | 697 | |||
BC038801.1 | - | 5 | 595 | |||
BC051345.1 | - | 5 | 646 | |||
BC110590.2 | - | 5 | 573 | 40027270 | ||
BX537420.1 | - | 5 | 602 | |||
D29956.1 | - | 5 | 721 |