Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALADL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005468.2 | NP_005459.2 | 17 | 2014 | ||
XM_011544706.2 | XP_011543008.1 | 15 | 2325 | |||
XM_011544707.2 | XP_011543009.1 | 18 | 1982 | |||
XM_011544708.2 | XP_011543010.1 | 15 | 2175 | |||
GenBank mRNA | AF010141.1 | - | 8 | 744 | ||
AJ012371.1 | - | 17 | 2014 | |||
AK311090.1 | - | 6 | 631 | |||
BG757994.1 | - | 5 | 490 |