Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005530.2 | NP_005521.1 | 6 | 566 | ||
XM_005254336.2 | XP_005254393.1 | 5 | 502 | |||
XM_017022118.1 | XP_016877607.1 | 5 | 581 | |||
XM_017022119.1 | XP_016877608.1 | 5 | 671 | |||
XM_017022120.1 | XP_016877609.1 | 4 | 466 | |||
GenBank mRNA | AK093531.1 | - | 4 | 1074 | ||
AK222973.1 | - | 6 | 550 | |||
AK296002.1 | - | 6 | 541 | |||
AK299970.1 | - | 6 | 478 | |||
AK315963.1 | - | 5 | 450 | |||
AK316051.1 | - | 5 | 438 | |||
AL110096.1 | - | 2 | 214 | |||
AL442090.1 | - | 3 | 488 | |||
BC021967.2 | - | 6 | 559 | 2989636 | ||
BC036227.1 | - | 6 | 567 | |||
CR749548.1 | - | 6 | 1230 | |||
U07681.1 | - | 6 | 544 |