Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008530.2 | NP_001008530.1 | 4 | 558 | ||
NM_005606.6 | NP_005597.3 | 3 | 440 | |||
XM_005267862.3 | XP_005267919.1 | 3 | 426 | |||
XM_005267863.3 | XP_005267920.1 | 3 | 426 | |||
XM_011536970.1 | XP_011535272.1 | 3 | 426 | |||
XM_017021463.1 | XP_016876952.1 | 3 | 250 | |||
XM_017021464.1 | XP_016876953.1 | 3 | 355 | |||
XM_017021465.1 | XP_016876954.1 | 3 | 244 | |||
XM_017021466.1 | XP_016876955.1 | 3 | 333 | |||
XM_017021467.1 | XP_016876956.1 | 3 | 344 | |||
GenBank mRNA | AK225822.1 | - | 3 | 331 | ||
AK291534.1 | - | 3 | 365 | |||
AK291560.1 | - | 3 | 365 | |||
AK297074.1 | - | 4 | 365 | |||
AK297780.1 | - | 4 | 297 | |||
BC003061.2 | - | 3 | 329 | 3504506 | ||
BX161380.1 | - | 3 | 346 | |||
BX161422.1 | - | 3 | 343 | |||
BX161434.1 | - | 4 | 468 | |||
CR457406.1 | - | 2 | 197 | |||
D55696.1 | - | 2 | 242 | |||
DA347449.1 | - | 3 | 440 | |||
Y09862.1 | - | 3 | 267 |