Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135099.1 | NP_001128571.1 | 10, 11 | 1239 | ||
NM_005656.3 | NP_005647.3 | 10, 11 | 1201 | |||
XM_005261043.2 | XP_005261100.1 | 9, 10 | 1081 | |||
XM_011529731.2 | XP_011528033.1 | 10, 11 | 1296 | |||
XM_011529732.2 | XP_011528034.1 | 9, 10 | 1123 | |||
XM_011529733.1 | XP_011528035.1 | 8, 9 | 900 | |||
GenBank mRNA | AF123453.1 | - | 8, 9 | 1067 | ||
AF270487.1 | - | 10, 11 | 1180 | |||
AF329454.1 | - | 10, 11 | 1194 | |||
AK222784.1 | - | 9, 10 | 1112 | |||
AK291813.1 | - | 10, 11 | 1156 | |||
AK296860.1 | - | 10, 11 | 1239 | |||
AK313338.1 | - | 11, 12 | 1501 | |||
BC035623.1 | - | 10, 11 | 1178 | |||
BC051839.1 | - | 10, 11 | 1172 | 6199625 | ||
U75329.1 | - | 9, 10 | 1123 |