Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256670.1 | NP_001243599.1 | 17 | 2819 | ||
NM_005700.4 | NP_005691.2 | 18 | 2909 | |||
NM_130443.3 | NP_569710.2 | 18 | 2912 | |||
GenBank mRNA | AJ271216.1 | - | 18 | 2508 | ||
AK021449.1 | - | 9 | 1131 | |||
AK025984.1 | - | 14 | 1917 | |||
AK222566.1 | - | 18 | 2486 | |||
AK222847.1 | - | 18 | 2487 | |||
AL137670.1 | - | 2 | 454 | |||
AL833475.1 | - | 18 | 2514 | |||
BC001446.2 | - | 17 | 2472 | 3138626 | ||
BC007221.1 | - | 18 | 2498 | 3349135 | ||
BC014038.2 | - | 18 | 2472 | 4547232 | ||
BC024271.1 | - | 17 | 2468 | 4564651 |