Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256670.1 | NP_001243599.1 | 17 | 2931 | ||
NM_005700.4 | NP_005691.2 | 18 | 3021 | |||
NM_130443.3 | NP_569710.2 | 18 | 3024 | |||
GenBank mRNA | AJ271216.1 | - | 18 | 2620 | ||
AK021449.1 | - | 9 | 1243 | |||
AK025984.1 | - | 14 | 2029 | |||
AK222566.1 | - | 18 | 2598 | |||
AL137670.1 | - | 2 | 566 | |||
AL833475.1 | - | 18 | 2626 | |||
BC001446.2 | - | 17 | 2584 | 3138626 | ||
BC007221.1 | - | 18 | 2610 | 3349135 | ||
BC014038.2 | - | 18 | 2584 | 4547232 | ||
BC024271.1 | - | 17 | 2580 | 4564651 |