Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174097.2 | NP_001167568.1 | 3 | 522 | ||
NM_001315537.1 | NP_001302466.1 | 3 | 300 | |||
NM_002300.7 | NP_002291.1 | 3 | 300 | |||
XM_006719074.2 | XP_006719137.1 | 3 | 416 | |||
GenBank mRNA | AK129881.1 | - | 3 | 251 | ||
AK312328.1 | - | 3 | 284 | |||
AL516910.3 | - | 3 | 521 | |||
AY189689.1 | - | 3 | 276 | |||
BC002362.1 | - | 3 | 267 | 2961445 | ||
BC015122.1 | - | 3 | 291 | 3685985 | ||
BC071860.1 | - | 3 | 232 | 4902461 | ||
BT019764.1 | - | 2 | 189 | |||
BT019765.1 | - | 2 | 189 | |||
BU540074.1 | - | 3 | 303 | |||
CX867875.1 | - | 3 | 497 | |||
EU794633.1 | - | 3 | 243 | |||
Y00711.1 | - | 3 | 243 |