Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006043.1 | NP_006034.1 | 2 | 1411 | ||
GenBank mRNA | AF105374.1 | - | 2 | 1411 | ||
AF105375.1 | - | 2 | 1411 | |||
AY359095.2 | - | 2 | 1774 |