Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252078.1 | NP_001239007.1 | 1, 2 | 169 | ||
NM_001252079.1 | NP_001239008.1 | 1, 2 | 169 | |||
NM_006313.2 | NP_006304.1 | 1, 2 | 169 | |||
XM_006719718.3 | XP_006719781.1 | 1, 2 | 167 | |||
XM_017020285.1 | XP_016875774.1 | 1, 2 | 120 | |||
GenBank mRNA | AB011101.2 | - | 1, 2 | 94 | ||
AF013990.1 | - | 1, 2 | 40 | |||
AF106069.1 | - | 1, 2 | 103 | |||
AF153604.1 | - | 1, 2 | 151 | |||
AK023703.1 | - | 1, 2 | 112 | |||
AK292337.1 | - | 1, 2 | 129 | |||
AK295947.1 | - | 1, 2 | 101 | |||
AK297759.1 | - | 1, 2 | 105 | |||
BC020688.1 | - | 1, 2 | 105 | |||
BC063454.1 | - | 1, 2 | 109 | |||
BC110908.1 | - | 1, 2 | 102 | |||
BC125123.1 | - | 1, 2 | 103 | 40118994 | ||
DB451322.1 | - | 1, 2 | 171 |