Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252078.1 | NP_001239007.1 | 9 | 1089 | ||
NM_006313.2 | NP_006304.1 | 8 | 1002 | |||
XM_005269261.2 | XP_005269318.1 | 3 | 293 | |||
XM_006719718.3 | XP_006719781.1 | 9 | 1087 | |||
XM_017020284.1 | XP_016875773.1 | 7 | 1721 | |||
XM_017020285.1 | XP_016875774.1 | 10 | 2426 | |||
GenBank mRNA | AB011101.2 | - | 8 | 927 | ||
AF013990.1 | - | 8 | 885 | |||
AF106069.1 | - | 8 | 936 | |||
AF153604.1 | - | 9 | 1071 | |||
BC125123.1 | - | 8 | 936 | 40118994 |