Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006373.3 | NP_006364.2 | 6 | 1846 | ||
GenBank mRNA | AK097072.1 | - | 7 | 1673 | ||
BC001913.1 | - | 6 | 1715 | |||
BC008725.2 | - | 6 | 1770 | 3162543 | ||
BC014279.2 | - | 6 | 1752 | 3834206 | ||
BC015041.2 | - | 6 | 1784 | 3919901 | ||
BC090858.1 | - | 6 | 1784 | 6528247 | ||
U25779.1 | - | 6 | 1454 |