Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001992.1 | NP_001001992.1 | 10 | 1183 | ||
NM_001032410.1 | NP_001027582.1 | 11 | 1249 | |||
NM_006447.2 | NP_006438.1 | 10 | 1186 | |||
XM_017028257.1 | XP_016883746.1 | 11 | 1333 | |||
XM_017028258.1 | XP_016883747.1 | 11 | 1212 | |||
XM_017028259.1 | XP_016883748.1 | 11 | 1184 | |||
XM_017028260.1 | XP_016883749.1 | 9 | 929 | |||
XM_017028261.1 | XP_016883750.1 | 11 | 1176 | |||
GenBank mRNA | AF113219.1 | - | 6 | 571 | ||
AF126736.1 | - | 10 | 1112 | |||
AK023247.1 | - | 5 | 535 | |||
AK025104.1 | - | 10 | 1149 | |||
AK025989.1 | - | 10 | 1152 | |||
AK222681.1 | - | 10 | 1115 | |||
AK222884.1 | - | 10 | 1108 | |||
AK294284.1 | - | 9 | 1047 | |||
AK302247.1 | - | 4 | 295 | |||
AY333928.1 | - | 10 | 1112 | |||
BC030777.2 | - | 10 | 1139 | 4824914 |