Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006536.5 | NP_006527.1 | 1 | 272 | ||
XM_011542448.2 | XP_011540750.1 | 1 | 214 | |||
GenBank mRNA | AB026833.1 | - | 1 | 151 | ||
AF043977.1 | - | 1 | 218 | |||
AF127980.1 | - | 1 | 110 | |||
AK290348.1 | - | 1 | 177 | |||
AK296719.1 | - | 1 | 215 | |||
AK307937.1 | - | 1 | 175 | |||
BC041096.1 | - | 1 | 173 | 5752653 |