Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006796.2 | NP_006787.2 | 8 | 1195 | ||
XM_011525601.2 | XP_011523903.1 | 8 | 1228 | |||
GenBank mRNA | AK129504.1 | - | 1 | 21 | ||
BC024282.1 | - | 9 | 1048 | |||
BC065016.1 | - | 8 | 1117 | 6169804 | ||
Y18314.1 | - | 8 | 1114 |