Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207048.1 | NP_001193977.1 | 8 | 2389 | ||
NM_001207049.1 | NP_001193978.1 | 8 | 2314 | |||
NM_012104.4 | NP_036236.1 | 9 | 3027 | |||
NM_138971.3 | NP_620427.1 | 9 | 2895 | |||
NM_138972.3 | NP_620428.1 | 9 | 2952 | |||
NM_138973.3 | NP_620429.1 | 9 | 2820 | |||
GenBank mRNA | AB032975.2 | - | 9 | 3009 | ||
AB177847.1 | - | 8 | 2251 | |||
AF200193.1 | - | 9 | 2525 | |||
AF201468.1 | - | 9 | 3020 | |||
AL833810.1 | - | 1 | 299 | |||
BC065492.1 | - | 9 | 3027 | 6538154 |