Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012105.4 | NP_036237.2 | 1, 2 | 783 | ||
NM_138991.2 | NP_620476.1 | 1, 2 | 783 | |||
NM_138992.2 | NP_620477.1 | 1, 2 | 783 | |||
XM_017028314.1 | XP_016883803.1 | 1, 2 | 455 | |||
GenBank mRNA | AF050171.1 | - | 1, 2 | 428 | ||
AF117892.1 | - | 1, 2 | 420 | |||
AF178532.1 | - | 1, 2 | 783 | |||
AF188276.1 | - | 1, 2 | 783 | |||
AF188277.1 | - | 1, 2 | 783 | |||
AF200192.1 | - | 1, 2 | 397 | |||
AF200342.1 | - | 1, 2 | 320 | |||
AF204944.1 | - | 1, 2 | 410 | |||
AF212252.1 | - | 1, 2 | 141 | |||
AK075539.1 | - | 2, 3 | 277 | |||
AK292056.1 | - | 1, 2 | 456 | |||
AY358927.1 | - | 1, 2 | 413 | |||
BC014453.1 | - | 1, 2 | 411 | 4868925 |