Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174146.1 | NP_001167617.1 | 3 | 513 | ||
NM_001174147.1 | NP_001167618.1 | 3 | 513 | |||
NM_002316.3 | NP_002307.2 | 3 | 513 | |||
GenBank mRNA | AF057135.1 | - | 3 | 437 | ||
BC069588.1 | - | 3 | 432 | |||
BC069601.1 | - | 3 | 437 | 7262383 | ||
BC112120.1 | - | 3 | 487 | 8327588 | ||
BC113491.1 | - | 3 | 487 | 8322543 | ||
BC143801.1 | - | 3 | 487 | 9052315 | ||
KU178166.1 | - | 3 | 437 | |||
KU178167.1 | - | 3 | 437 | |||
U77457.1 | - | 1 | 155 |